Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA Mouse anti-Mammals, Clone: 5A9, MilliporeSigma™

Catalog No. MAB316MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB316MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB316MI Supplier MilliporeSigma Supplier No. MAB316
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-GABA Antibody, clone 5A9 detects level of GABA and has been published and validated for use in ELISA, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABA
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 5A9
Conjugate Unconjugated
Formulation Purified mouse monoclonal in PBS, pH 7.4, 1% BSA, and 0.1% sodium azide as a preservative.
Host Species Mouse
Immunogen GABA-coupled to BSA with glutaraldehyde.
Purification Method Protein A purfied
Quantity 50 μg
Primary or Secondary Primary
Content And Storage 2-8°C for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.