Learn More
GABA (No Gluteraldehyde) Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5016
Description
This Anti-GABA (No Gluteraldehyde) Antibody is validated for use in immunohistochemistry for the detection of GABA (No Gluteraldehyde).
Specifications
GABA (No Gluteraldehyde) | |
Polyclonal | |
Purified immunoglobulin. Liquid in stabilizing buffer (Phosphate buffer with sterile goat serum) containing 0.05% thimerosal. | |
GABA-glutaraldehyde | |
Primary | |
2-8°C for up to 6 months, do not freeze |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
500 μL | |
Rat | |