Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA Transporter-2 Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB1572MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1572MI 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1572MI Supplier MilliporeSigma Supplier No. AB1572
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-GABA Transporter-2 Antibody is validated for use in immunohistochemistry, western blotting for the detection of GABA Transporter-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABA Transporter-2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified antibody. Purified in PBS with 0.1% sodium azide.
Gene Symbols SLC6A13; GAT-2; GAT2
Host Species Rabbit
Immunogen GAT-2, C-terminus (aa 593-602)
Purification Method Affinity Purified
Quantity 10 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_016615.2
Target Species Mouse, Rat
Content And Storage Lyophilized: 2-8°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.