Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-GADD45α, Polyclonal,

Catalog No. 071230MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-123-0MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-123-0MI Supplier MilliporeSigma™ Supplier No. 071230

Specifications

Antigen GADD45α
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in buffer containing 0.02M Phosphate with 0.15M NaCl, pH 7.6 and 0.1% sodium azide as a preservative.
Gene Symbols GADD45A; DDIT1; GADD45; DDIT-1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the N-terminus of human GADD45α (MTLEEFSAGEQKTERMDKVGDALEE).
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001924
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquotes for up to 6 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.