Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GADD45A, Mouse, Clone: 3D12, Abnova™

Catalog No. 89021280 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-280 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-280 Supplier Abnova Corporation Supplier No. H00001647M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GADD45A.

This gene is a member of a group of genes whose transcript levels are increased following stressful growth arrest conditions and treatment with DNA-damaging agents. The protein encoded by this gene responds to environmental stresses by mediating activation of the p38/JNK pathway via MTK1/MEKK4 kinase. The DNA damage-induced transcription of this gene is mediated by both p53-dependent and -independent mechanisms. [provided by RefSeq

Sequence: TLIQAFCCENDINILRVSNPGRLAELLLLETDAGPAASEGAEQPPDLHCVLVTNPHSSQWKDPALSQLICFCRESRYMDQWVPVINLPER

Specifications

Antigen GADD45A
Applications ELISA, In situ PLA, Western Blot
Classification Monoclonal
Clone 3D12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GADD45A.
Formulation PBS with no preservative; pH 7.4
Gene GADD45A
Gene Accession No. BC011757
Gene Alias DDIT1/GADD45
Gene Symbols GADD45A
Host Species Mouse
Immunogen GADD45A (AAH11757, 76 a.a. ∼ 165 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1647
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.