Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV1 Gag, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Supplier:  Thermo Scientific PA127932

Catalog No. PIPA127932


Description

Description

Matrix protein p17 targets Gag and Gag-Pol polyproteinss to the plasma membrane via a multipartite membrane binding signal. that includes its myristoylated N-terminus. Also mediates nuclear localization of the preintegration complex. Implicated in the release from host cell mediated by Vpu.

PA1-27932 detects gag from HIV infected samples.PA1-27932 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27526 immunogen is fragment of Gag polyprotein.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HIV1 Gag
Polyclonal
Human immunodeficiency virus, HIV-1
Fragment of Gag polyprotein.
50 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Purified
Western Blot
Unconjugated
Chicken
Antigen affinity chromatography
RUO
Virus
Antibody
IgY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.