Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

meiotic nuclear divisions 1 homolog (S. cerevisiae), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89008306 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-306 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-306 Supplier Abnova Corporation Supplier No. H00084057B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human GAJ protein.

The product of the MND1 gene associates with HOP2 (MIM 608665) to form a stable heterodimeric complex that binds DNA and stimulates the recombinase activity of RAD51 (MIM 179617) and DMC1 (MIM 602721) (Chi et al., 2007 [PubMed 17639080]). Both the MND1 and HOP2 genes are indispensable for meiotic recombination.[supplied by OMIM

Sequence: MSKKKGLSAEEKRTRMMEIFSETKDVFQLKDLEKIAPKEKGITAMSVKEVLQSLVDDGMVDCERIGTSNYYWAFPSKALHARKHKLEVLESQLSEGSQKHASLQKSIEKAKIGRCETEERTRLAKELSSLRDQREQLKAEVEKYKDCDPQVVEEIRQANKVAKEAANRWTDNIFAIKSWAKRKFGFEENKIDRTFGIPEDFDYID

Specifications

Antigen meiotic nuclear divisions 1 homolog (S. cerevisiae)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human GAJ protein.
Formulation No additive
Gene MND1
Gene Accession No. BC032142
Gene Alias GAJ
Gene Symbols MND1
Host Species Mouse
Immunogen GAJ (AAH32142, 1 a.a. ∼ 205 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 84057
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.