Learn More
GAK, Mouse, Clone: 4C10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant GAK.
Supplier: Abnova Corporation H00002580M01
Description
In all eukaryotes, the cell cycle is governed by cyclin-dependent protein kinases (CDKs), whose activities are regulated by cyclins and CDK inhibitors in a diverse array of mechanisms that involve the control of phosphorylation and dephosphorylation of Ser, Thr or Tyr residues. Cyclins are molecules that possess a consensus domain called the 'cyclin box.' In mammalian cells, 9 cyclin species have been identified, and they are referred to as cyclins A through I. Cyclin G is a direct transcriptional target of the p53 tumor suppressor gene product and thus functions downstream of p53. GAK is an association partner of cyclin G and CDK5. [provided by RefSeq]
Sequence: SAVAPTPATEGPLFSPGGQPAPCGSQASWTKSQNPDPFADLGDLSSGLQGSPAGFPPGGFIPKTATTPKGSSSWQTSRPPAQGASWPPQAKPPPKACTQPSpecifications
GAK | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC008668 | |
GAK | |
GAK (AAH08668, 151 a.a. ∼ 250 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Immunoprecipitation, Western Blot | |
4C10 | |
Mouse monoclonal antibody raised against a partial recombinant GAK. | |
GAK | |
FLJ16629/FLJ40395/MGC99654 | |
Mouse | |
Affinity chromatography | |
RUO | |
2580 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.