Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-GAL4 (DNA binding domain), Polyclonal,

Catalog No. 06262MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-262-MI 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-262-MI Supplier MilliporeSigma™ Supplier No. 06262

Specifications

Antigen GAL4 (DNA binding domain)
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide as a preservative.
Host Species Rabbit
Immunogen The GAL4 amino terminal fragment of yeast, containing only the DNA-binding domain (1-147) expressed in E. coli
Purification Method Ammonium sulfate precipitation
Quantity 250 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_015076
Target Species S. cerevisiae, Yeast
Content And Storage −20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.