Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galactocerebroside Mouse anti-Chicken, Bovine, Human, Mouse, Rat, Rabbit, Clone: mGalC, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB342
Description
Anti-Galactocerebroside Antibody, clone mGalC is an antibody against Galactocerebroside for use in ELISA, immunocytochemistry, and immunohistochemistry.
Specifications
| Galactocerebroside | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| Protein A purfied | |
| Primary | |
| −20°C for up to 12 months. | |
| IgG3 |
| ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
| mGalC | |
| Purified mouse monoclonal in buffer containing 10mM Potassium Phosphate, 150mM NaCl, pH 7.4 containing 0.09% sodium azide. | |
| Synaptic plasma membranes from bovine hippocampus. Ranscht, B. et al. PNAS 79(8):2709-2713 (1982) | |
| 50 μg | |
| Bovine, Chicken, Human, Mouse, Rabbit, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction