Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galactocerebroside Mouse anti-Chicken, Bovine, Human, Mouse, Rat, Rabbit, FITC, Clone: mGalC, MilliporeSigma™

Catalog No. FCMAB312FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB312FMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB312FMI Supplier MilliporeSigma Supplier No. FCMAB312F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Galactocerebroside using this Milli-Mark™ Anti-Galactocerebroside-FITC Antibody, clone mGalC validated for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galactocerebroside
Applications Flow Cytometry
Classification Monoclonal
Clone mGalC
Conjugate FITC
Formulation Purified mouse monoclonal IgG3 conjugated to FITC in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Accession No. NP_001032614
Gene Symbols GALC, EC 3.2.1.46
Host Species Mouse
Immunogen Synaptic plasma membranes from bovine hippocampus.
Purification Method Protein A Purified
Quantity 100 Tests
Primary or Secondary Primary
Target Species Bovine, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from Light.
Form Purified
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.