Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Β Galactosidase Rabbit anti-Vertebrates, E. coli, Polyclonal, MilliporeSigma™

Catalog No. AB986MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB986MI 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB986MI Supplier MilliporeSigma Supplier No. AB986
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-β Galactosidase Antibody, bacterial detects level of β Galactosidase and has been published and validated for use in ELISA, immunoprecipitation, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen β Galactosidase
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit antisera prepared by delipidation and defibrination. Lyophilized. Reconstitute with 2 mL of sterile deionized water. Contains 0.02M Potassium Phosphate, 0.15M NaCl and 0.01% sodium azide after reconstitution.
Gene Symbols GLB1; Lactase; S-Gal; ELNR1; EBP; EC 3.2.1.23
Host Species Rabbit
Immunogen Beta galactosidase from E-coli.
Quantity 2 mL
Primary or Secondary Primary
Gene ID (Entrez) NM_000404.2; NM_001079811.1
Target Species E. coli, Vertebrates
Content And Storage Lyophilized: 2-8°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.