Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galanin Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB2233MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2233MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2233MI Supplier MilliporeSigma Supplier No. AB2233
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Galanin using this Anti-Galanin Antibody validated for use in immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galanin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Serum with 0.05% NaN3
Gene Symbols GAL1 ; GALN; GLNN ; GMAP; MGC40167; galanin
Host Species Rabbit
Immunogen Synthetic Linear Peptide
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_057057
Target Species Rat
Content And Storage −20°C for 12 months
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.