Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galanin Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5909
Description
This Anti-Galanin Antibody is validated for use in ELISA, immunohistochemistry for the detection of Galanin.
Specifications
Galanin | |
Polyclonal | |
Rabbit serum. Liquid with 0.05% sodium azide. | |
Rabbit | |
50 μL | |
NM_015973.3 | |
−20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
ELISA, Immunohistochemistry | |
Unconjugated | |
GAL; GAL1; galanin; GALN; GLNN; GMAP; MGC40167 | |
Synthetic peptide from human Galanin. The immunogen peptide is available for use in absorbtion studies (Catalog Number AG370). | |
Primary | |
Human | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction