Learn More
Description
This gene encodes a lysosomal protein which hydrolyzes the galactose ester bonds of galactosylceramide, galactosylsphingosine, lactosylceramide, and monogalactosyldiglyceride. Mutations in this gene have been associated with Krabbe disease, also known as globoid cell leukodystrophy. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Specifications
Specifications
| Antigen | GALC |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2D1 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full-length recombinant GALC. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | GALC |
| Gene Accession No. | BC036518 |
| Gene Symbols | GALC |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.