Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin-3 Mouse anti-Human, Unlabeled, Clone: B2C10, BD

Catalog No. BDB556904 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556904 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556904 Supplier BD Biosciences Supplier No. 556904
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with Galectin-3, a 30kDa protein, a member of a newly defined family of β-galactoside-binding animal lectins. It has been previously referred to as IgE-binding protein (εBP) and carbohydrate-binding protein 35 (CBP35). It is expressed on many tissues and cell types. Galectin-3 can be found in the nucleus and/or cytoplasm, on the cell surface and in the extra cellular matrix. Its function has been found to be diverse. It has been reported to be markedly elevated in the nucleus of proliferating cells, suggesting that Galectin-3 may play a role in cell growth regulation. Studies of Galectin-3 on the cell surface have shown it to be a non-integrin, laminin-binding protein, suggesting its role in cell adhesion to basement membranes. Monoclonal antibody B2C10 is suitable for immunohistochemical staining of parafin-embedded tissue sections.

Host Species: Mouse
Clone: B2C10
Isotype: IgG1, κ
Cross Reactivity: Human

Immunohistochemistry, Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin-3
Applications Flow Cytometry
Classification Monoclonal
Clone B2C10
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human Galectin-3
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.