Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin-3 Rat anti-Human, Mouse, Clone: M3/38, MilliporeSigma™

Catalog No. MABT51MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT51MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT51MI Supplier MilliporeSigma Supplier No. MABT51
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Galectin-3 Antibody, clone M3/38 is an antibody against Galectin-3 for use in western blot, immunoprecipitation, immunohistochemistry and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin-3
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone M3/38
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001139425
Gene Symbols LGALS3, L-31, GAL3, Galectin-3, GALIG, MAC2, GALBP, LGALS2, MAC-2, CBP35
Host Species Rat
Immunogen Plasma membrane glycoproteins corresponding to mouse Galectin-3.
Purification Method Protein G Purified
Quantity 50 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.