Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Galectin-9 Clone: 1G3,

Catalog No. MABT833MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABT833MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT833MI Supplier MilliporeSigma™ Supplier No. MABT833
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Galectin-9 Clone: 1G3 in Human samples, and it is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting

Galectin-9 (UniProt O00182; also known as Ecalectin, Gal-9, Tumor antigen HOM-HD-21, Urate transporter/channel protein) is encoded by the LGALS9 (also known as HUAT, LGALS9A) gene (Gene ID 3965) in human. The beta-galactoside-binding lectin, Galectin-9, possesses two distinct carbohydrate recognition domains (CRDs) linked together by a peptide domain of different length among the S, M, and L spliced isoforms. Galectin-9 plays a key role in a negative feed-back mechanism against Th1 immune response, where galectin-9 production from various cell types (e.g. fibroblasts and endothelial cells) is induced by interferon-gamma produced by CD4+ Th1 lymphocytes. The up-regulated galectin-9 in turn suppresses CD4+ Th1 lymphocytes, at least in part through stimulation of the Tim-3 receptor. The Tim-3 receptor on CD4+ Th1 cells from patients with multiple sclerosis (MS), rheumatoid arthritis, and auto-immune hepatitis is defective in its response to galectin-9. In addition, excessive galectin-9 production is reported in two human diseases associated with oncogenic viruses, nasopharyngeal carcinomas (NPC) associated with the Epstein-Barr virus (EBV) and chronic infection by the hepatitis C virus (HCV).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin-9
Applications ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1G3
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. O00182
Gene Symbols LGALS9
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human Galectin-9.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_033665
Test Specificity Clone 1G3 reacts with all three (L, M, and S) galectin-9 isoforms, but not galectin 1, 2, 3, 4, 7, 8, or 10.
Target Species Human
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.