Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

galactokinase 2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89005414 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-005-414 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-005-414 Supplier Abnova Corporation Supplier No. H00002585B02
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human GALK2 protein.

This gene encodes a highly efficient N-acetylgalactosamine (GalNAc) kinase, which has galactokinase activity when galactose is present at high concentrations. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MATESPATRRVQVAEHPRLLKLKEMFNSKFGSIPKFYVRAPGRVNIIGEHIDYCGYSVLPMAVEQDVLIAVEPVKTYALQLANTNPLYPDFSTSANNIQIDKTKPLWHNYFLCGLKGIQEHFGLSNLTGMNCLVDGNIPPSSGLSSSSALVCCAGLVTLTVLGRNLSKVELAEICAKSERYIGTEGGGMDQSISFLAEEGTAKLIEFSPLRATDVKLPSGAVFVIANSCVEMNKAATSHFNIRVMECRLAAKLLAKYKSLQWDKVLRLEEVQAKLGISLEEMLLVTEDALHPEPYNPEEICRCLGISLEELRTQILSPNTQDVLIFKLYQRAKHVYSEAARVLQFKKICEEAPENMVQLLGELMNQSHMSCRDMYECSCPELDQLVDICRKFGAQGSRLTGAGWGGCTVSMVPADKLPSFLANVHKAYYQRSDGSLAPEKQSLFATKPGGGALVLLEA

Specifications

Antigen galactokinase 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human GALK2 protein.
Formulation No additive
Gene GALK2
Gene Accession No. NM_002044.2
Gene Alias GK2/MGC1745
Gene Symbols GALK2
Host Species Mouse
Immunogen GALK2 (NP_002035.1, 1 a.a. ∼ 458 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 2585
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.