Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gamma-synuclein (SNCG) Mouse anti-Human, Clone: 3D2.1, MilliporeSigma™

Catalog No. MABN314MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN314MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN314MI Supplier MilliporeSigma Supplier No. MABN314
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Gamma-synuclein (SNCG), clone 3D2.1 Antibody is validated for use in Western Blotting for the detection of Gamma-synuclein (SNCG).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gamma-synuclein (SNCG)
Applications Western Blot
Classification Monoclonal
Clone 3D2.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_003078
Gene Symbols SNCG, BCSG1, PERSYN, PRSN
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human Gamma-synuclein (SNCG).
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.