Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAMT, Mouse, Clone: 3H4, Abnova™

Catalog No. 89022265 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-265 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-265 Supplier Abnova Corporation Supplier No. H00002593M04
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GAMT.

The protein encoded by this gene is a methyltransferase that converts guanidoacetate to creatine, using S-adenosylmethionine as the methyl donor. Defects in this gene have been implicated in neurologic syndromes and muscular hypotonia, probably due to creatine deficiency and accumulation of guanidinoacetate in the brain of affected individuals. Two transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq

Sequence: PLSEETWHTHQFNFIKNHAFRLLKPGGVLTYCNLTSWGELMKSKYSDITIMFEETQVPALLEAGFRRENIRTEVMALVPPADCRYYAFPQMITPLVTK

Specifications

Antigen GAMT
Applications ELISA
Classification Monoclonal
Clone 3H4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GAMT.
Formulation PBS with no preservative; pH 7.4
Gene GAMT
Gene Accession No. NM_000156
Gene Alias PIG2/TP53I2
Gene Symbols GAMT
Host Species Mouse
Immunogen GAMT (NP_000147.1, 138 a.a. ∼ 235 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2593
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.