Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Ganglioside GM1, Asialo, Polyclonal

Catalog No. 345758100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
34-575-8100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-575-8100UL Supplier MilliporeSigma™ Supplier No. 345758100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Ganglioside GM1, Asialo Clone: in samples, and it is validated for ELISA, Immuno Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ganglioside GM1, Asialo
Applications ELISA, Immunoblot, Immunoblot
Classification Polyclonal
Conjugate Unconjugated
Formulation Undiluted serum.
Host Species Rabbit
Immunogen purified, bovine brain asialo-ganglioside GM1
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes ganglioside GM1 asialo.
Content And Storage −20°C
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.