Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAP-43/Neuromodulin Mouse, Unlabeled, Clone: 31, BD

Catalog No. BDB612262 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612262 50 μg
BDB612263 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612262 Supplier BD Biosciences Supplier No. 612262

Mouse Monoclonal Antibody

Neuromodulin (GAP-43, B-50, F-1) is a neural-specific protein that represents about one percent of the total membrane protein in growth cones from developing rat brain. Growth cones are located at the distal tip of neuronal processes and are critical for transduction of extracellular signals into directed growth. Neuromodulin is attached to the membranes of the neuronal growth cones during development. In the adult, it is attached to the presynaptic membranes in discrete subsets of synapses. Neuromodulin can bind to calmodulin, as well as inhibit phosphatidylinositol phosphate kinase (PIP kinase) activity. It is also a prominent substrate of Protein Kinase C (PKC) and may mediate some of the effects of PKC on growth cone function. As may be expected, expression and phosphorylation of neuromodulin are closely associated with axonal outgrowth and repair. Phosphorylation of neuromodulin by PKC has been correlated with the persistence of long term potentiation and the translocation of PKC to synaptosomal membranes. This PKC-mediated phosphorylation does not appear to require neuromodulin membrane association. It has also been demonstrated that the GTP-binding protein, G[o], is a main component of growth cone membranes. The binding of GTP--S to G[o] is stimulated by neuromodulin and this activity resides within the amino-terminal region of the protein. This region of neuromodulin shares sequence homology with the cytoplasmic tails of G-linked transmembrane receptors, suggesting the presence of one binding site on G[o] for both neuromodulin and transmembrane receptors.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen GAP-43/Neuromodulin
Applications Western Blot
Classification Monoclonal
Clone 31
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human GAP-43/Neuromodulin aa. 1-144
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.