Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GAP43 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA124970
Description
PA1-24970 detects GAP-43 from human, mouse, rat, feline, and monkey samples.
GAP43 (Neuromodulin) is associated with nerve growth. GAP43 is expressed by developing and regenerating neurons, and to a lesser extent reactive glial cells. It is a major component of the motile growth cones that form the tips of elongating axons. GAP43 also plays a role in axonal and dendritic filopodia induction.
Specifications
GAP43 | |
Polyclonal | |
Unconjugated | |
GAP43 | |
Axonal membrane protein GAP-43; B-50; Basp2; brain abundant, membrane attached signal protein 2; calmodulin-binding protein P-57; Gap43; GAP-43; growth accentuating protein 43; growth associated protein 43; growth-associated protein 43; MGC156666; nerve growth-related peptide GAP43; neural phosphoprotein B-50; Neuromodulin; neuron growth-associated protein 43; neuron protein F1; PP46; Protein F1 | |
Rabbit | |
50 μL | |
Primary | |
Human, Mouse, Rat, Monkey, Feline | |
Antibody | |
IgG |
Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot, Immunocytochemistry | |
Conc. Not Determined | |
whole serum with 0.02% sodium azide | |
P06837, P07936, P17677, Q6S9D9 | |
GAP43 | |
Purified GAP43 from cat brain. | |
RUO | |
14432, 2596, 29423, 493873 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction