Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAPDH Chicken anti-Human, Mouse, Rat, Horse, Canine, Polyclonal, MilliporeSigma™
Chicken Polyclonal Antibody
Supplier: MilliporeSigma AB2302
Description
Anti-GAPDH Antibody detects level of GAPDH and has been published and validated for use in western blotting.
Specifications
GAPDH | |
Polyclonal | |
Purified chicken polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
Chicken | |
Affinity Purified | |
Primary | |
Canine, Equine, Human, Mouse, Rat |
Western Blot | |
Unconjugated | |
GAPDH; G3PDH; GAPD; G3PD | |
KLH-conjugated linear peptide corresponding to the WARS interacting region of human GAPDH. | |
100 μg | |
NP_002037 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction