Learn More
GAPDH, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human GAPDH peptide using ARM Technology.
Supplier: Abnova Corporation H00002597K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The product of this gene catalyzes an important energy-yielding step in carbohydrate metabolism, the reversible oxidative phosphorylation of glyceraldehyde-3-phosphate in the presence of inorganic phosphate and nicotinamide adenine dinucleotide (NAD). The enzyme exists as a tetramer of identical chains. Many pseudogenes similar to this locus are present in the human genome. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
GAPDH | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
GAPDH | |
GAPDH | |
A synthetic peptide of human GAPDH | |
RUO | |
2597 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
glyceraldehyde-3-phosphate dehydrogenase | |
GAPDH | |
G3PD, GAPD, MGC88685 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.