Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GARP Mouse anti-Human, APC, Clone: 7B11 (also known as CMSSC-7B11), BD

Catalog No. BDB562341 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562341 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562341 Supplier BD Biosciences Supplier No. 562341
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 7B11 (also known as CMSSC-7B11) monoclonal antibody specifically binds to human GARP (Glycoprotein A repetitions predominant). The LRRC32 (Leucine rich repeat containing 32) gene encodes the 662 amino acid-residue, 80 kDa transmembrane GARP glycoprotein that has an extracellular region composed primarily of 20 leucine-rich repeats. GARP is specifically expressed on Treg cells activated through the T cell receptor (TCR). Ectopic expression of GARP in human naοve T cells inhibited their proliferation and cytokine secretion upon TCR activation. Remarkably, GARP over-expression in naοve T cells induced expression of FoxP3 and endowed them with a partial suppressive function. The extracellular, but not the cytoplasmic region, of GARP was necessary for these functions. GARP serves as a receptor for latent TGF-beta which may play a role in the suppressive action of Treg cells. GARP is also expressed on platelets and other tissues, however the function on these cells is not known.

TRUSTED_SUSTAINABILITY

Specifications

Antigen GARP
Applications Flow Cytometry
Classification Monoclonal
Clone 7B11 (also known as CMSSC-7B11)
Conjugate APC
Description LRRC32; Garpin; Glycoprotein A repetitions predominant
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human LRRC32
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.