Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GARS Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA515148
Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
GARS is a glycyl-tRNA synthetase, one of the aminoacyl-tRNA synthetases that charge tRNAs with their cognate amino acids. This protein is an (alpha)2 dimer which belongs to the class II family of tRNA synthetases. The protein has been shown to be a target of autoantibodies in the human autoimmune diseases, polymyositis or dermatomyositis.
Specifications
GARS | |
Polyclonal | |
GARS | |
Glycyl-tRNA synthetase; Glycyl-tRNA synthetase; Glycine--tRNA ligase; GlyRS; GARS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2617 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
P41250 | |
GARS | |
KLH conjugated synthetic peptide between 706-739 amino acids from the C-terminal region of human GARS | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction