Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA-4 Mouse anti-Mouse, Human, Clone: 13G4.2, MilliporeSigma™

Catalog No. MAB4344MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4344MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4344MI Supplier MilliporeSigma Supplier No. MAB4344

Mouse Monoclonal Antibody

Anti-GATA-4 Antibody, clone 13G4.2 detects level of GATA-4 and has been published and validated for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GATA-4
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 13G4.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_002043
Gene Symbols GATA4, GATA-4
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human GATA-4.
Purification Method Protein G Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.