Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA binding protein 2, Mouse, Clone: 1G5, Abnova™

Catalog No. 89000676 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-676 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-676 Supplier Abnova Corporation Supplier No. H00002624M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GATA2.

This gene encodes a member of the GATA family of zinc-finger transcription factors that are named for the consensus nucleotide sequence they bind in the promoter regions of target genes. The encoded protein plays an essential role in regulating transcription of genes involved in the development and proliferation of hematopoietic and endocrine cell lineages. Alternative splicing results in multiple transcript variants

Sequence: MEVAPEQPRWMAHPAVLNAQHPDSHHPGLAHNYMEPAQLLPPDEVDVFFNHLDSQGNPYYANPAHARARVSYSPAHARLTGGQMCRPHLLHSPGLPWLDGGK

Specifications

Antigen GATA binding protein 2
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1G5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GATA2.
Formulation PBS with no preservative; pH 7.4
Gene GATA2
Gene Accession No. BC018988
Gene Alias MGC2306/NFE1B
Gene Symbols GATA2
Host Species Mouse
Immunogen GATA2 (AAH18988, 1 a.a. ∼ 102 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 2624
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.