Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA3, Mouse, Clone: 3A4, Abnova™

Catalog No. 89022274 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-274 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-274 Supplier Abnova Corporation Supplier No. H00002625M09
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GATA3.

Sequence: LGSHHTASPWNLSPFSKTSIHHGSPGPLSVYPPASSSSLSGGHASPHLFTFPPTPPKDVSPDPSLSTPGSAGSARQDEKECLKYQVPLPDSMKLESSH

Specifications

Antigen GATA3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3A4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GATA3.
Formulation PBS with no preservative; pH 7.4
Gene GATA3
Gene Accession No. NM_001002295
Gene Alias HDR/MGC2346/MGC5199/MGC5445
Gene Symbols GATA3
Host Species Mouse
Immunogen GATA3 (NP_001002295.1, 103 a.a. ∼ 200 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2625
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.