Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA4 Mouse, Alexa Fluor 488, Clone: L97-56, BD

Catalog No. BDB560330 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560330 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560330 Supplier BD Biosciences Supplier No. 560330
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The L97-56 monoclonal antibody reacts with GATA4 (GATA-binding protein 4), a member of the GATA family of zinc finger-containing transcription factors that bind to the GATA nucleotide sequence. This ∽50kDa nuclear protein is expressed in mesodermal and definitive endodermal tissues such as the gastrointestinal tract, gonads, and heart. Genetic studies suggest that GATA4 regulates embryonic cardiac development: in mice, disruption of the GATA4 gene leads to defects in heart tube formation, while mutations of GATA4 are associated with atrial septal defects in humans. In the adult heart, GATA4 regulates differentiated gene expression. The roles of GATA4 in endocrine and reproductive functions were recently reviewed.

Host Species: Mouse
Clone: L97-56
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human GATA4 Peptide

Bioimaging, Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen GATA4
Applications Flow Cytometry
Classification Monoclonal
Clone L97-56
Conjugate Alexa Fluor 488
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human GATA4 Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.