Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA zinc finger domain containing 2A, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004656 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-656 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-656 Supplier Abnova Corporation Supplier No. H00054815A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant GATAD2A.

Sequence: QIQKEATAQKPTGSVGSTVTTPPPLVRGTQNIPAGKPSLQTSSARMPGSVIPPPLVRGGQQASSKLGPQASSQVVMPPLVRGAQQIHSIRQHSSTGPPPLLLAPRASVP

Specifications

Antigen GATA zinc finger domain containing 2A
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant GATAD2A.
Formulation 50% glycerol
Gene GATAD2A
Gene Accession No. NM_017660
Gene Alias FLJ20085/FLJ21017/p66alpha
Gene Symbols GATAD2A
Host Species Mouse
Immunogen GATAD2A (NP_060130, 26 a.a. ∼ 134 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 54815
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.