Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GBA, Mouse, Clone: 2H4, Abnova™

Catalog No. 89022276 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-276 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-276 Supplier Abnova Corporation Supplier No. H00002629M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GBA.

This gene encodes a lysosomal membrane protein that cleaves the beta-glucosidic linkage of glycosylceramide, an intermediate in glycolipid metabolism. Mutations in this gene cause Gaucher disease, a lysosomal storage disease characterized by an accumulation of glucocerebrosides. A related pseudogene is approximately 12kb downstream of this gene on chromosome 1. Alternative splicing results in multiple transcript variants encoding the same protein. [provided by RefSeq

Sequence: SYFSEEGIGYNIIRVPMASCDFSIRTYTYADTPDDFQLHNFSLPEEDTKLKIPLIHRALQLAQRPVSLLASPWTSPTWLKTNGAVNGKGS

Specifications

Antigen GBA
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2H4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GBA.
Formulation PBS with no preservative; pH 7.4
Gene GBA
Gene Accession No. NM_000157
Gene Alias GBA1/GCB/GLUC
Gene Symbols GBA
Host Species Mouse
Immunogen GBA (NP_000148, 146 a.a. ∼ 235 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2629
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.