Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GBF1 (Golgi-specific Brefeldin A (BFA)-resistance Factor 1) Mouse, Unlabeled, Clone: 25, BD

Catalog No. BDB612116 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612116 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612116 Supplier BD Biosciences Supplier No. 612116
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Golgi apparatus is a complex and dynamic organelle that functions in protein sorting and modification. Numerous structural and regulatory proteins are involved in the budding, docking, and fusion of Golgi-directed vesicles. Golgi-specific Brefeldin A (BFA)-resistance Factor (GBF1) is a guanine nucleotide exchange factor that contains a centrally located Sec7 domain, and a proline-rich region at the C-terminus. The Sec7 domain is commonly found in a variety of secretory proteins, and is required for activation of ARFs. GBF1 mRNA is ubiquitously expressed, and GBF1 protein localizes to the Golgi, as well as to perinuclear structures that contain COPI. BFA is a fungal heterocyclic lactone that disrupts membrane recruitment of ARFs. This action blocks protein secretion, and changes the morphology of various organelles, such as the Golgi. Overexpression of GBF1 suppresses BFA-induced changes in Golgi morphology, ARF activation, and coat protein recruitment. GBF1 interacts preferentially with ARF5 in the presence of Mg2+;. Thus, GBF1 is a Golgi-specific guanine nucleotide exchange factor that activates ARFs through a mechanism that interferes with BFA action.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen GBF1 (Golgi-specific Brefeldin A (BFA)-resistance Factor 1)
Applications Western Blot
Classification Monoclonal
Clone 25
Concentration 250μg/mL
Conjugate Unconjugated
Description Golgi-specific Brefeldin A (BFA)-resistance Factor 1
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human GBF1 aa. 1266-1379
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.