Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GBGT1, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89202144 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-202-144 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-202-144 Supplier Abnova Corporation Supplier No. H00026301K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human GBGT1 peptide using ARM Technology.

This gene encodes a member of the histo-blood group ABO gene family that encodes glycosyltransferases with related but distinct substrate specificity. This protein plays a role in synthesizing Forssman glycolipid (FG), a member of the globoseries glycolipid family. Human cells do not normally produce FG but produce the precursor glycolipids globotriaosylceramide and globoside. This protein may be involved in the tropism and binding of pathogenic organisms. [provided by RefSeq

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen GBGT1
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description globoside alpha-1,3-N-acetylgalactosaminyltransferase 1
Formulation In 1X PBS, pH 7.4
Gene GBGT1
Gene Accession No. GBGT1
Gene Alias A3GALNT, FS, MGC44848, UNQ2513
Gene Symbols GBGT1
Host Species Rabbit
Immunogen A synthetic peptide of human GBGT1
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26301
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.