Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gC1q-R/p33 Mouse anti-Human, Mouse, Clone: 74.5.2, MilliporeSigma™

Catalog No. MAB1160MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1160MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1160MI Supplier MilliporeSigma Supplier No. MAB1160
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-gC1q-R/p33 Antibody, C-terminus, aa 204-218, clone 74.5.2 is validated for use in ELISA, flow cytometry, immunoprecipitation, western blotting for the detection of gC1q-R/p33.
TRUSTED_SUSTAINABILITY

Specifications

Antigen gC1q-R/p33
Applications ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 74.5.2
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.02M PBS pH 7.6, 0.25M NaCl with 0.1% sodium azide.
Gene Accession No. NM_012072.3
Gene Symbols CD93, MXRA4, C1qRp, C1qR(P), C1qR, dJ737E23.1, C1QR1, CDw93, p, C1qRP
Host Species Mouse
Immunogen Recombinant human gC1q-R.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.