Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

germinal center expressed transcript 2, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016636 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-636 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-636 Supplier Abnova Corporation Supplier No. H00257144B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human GCET2 protein.

This gene encodes a protein which may function in signal transduction pathways and whose expression is elevated in germinal cell lymphomas. It contains a putative PDZ-interacting domain, an immunoreceptor tyrosine-based activation motif (ITAM), and two putative SH2 binding sites. In B cells, its expression is specifically induced by interleukin-4. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq

Sequence: MGNSLLRENRRQQNTQEMPWNVRMQSPKQRTSRCWDHHIAEGCFCLPWKKILIFEKRQDSQNENERMSSTPIQDNVDQTYSEELCYTLINHRVLCTRPSGNSAEEYYENVPCKAERPRESLGGTETEYSLLHMPSTDPRHARSPEDEYELLMPHRISSHFLQQPRPLMAPSETQFSHL

Specifications

Antigen germinal center expressed transcript 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human GCET2 protein.
Formulation PBS with no preservative; pH 7.4
Gene GCET2
Gene Accession No. NM_152785.3
Gene Alias GCAT2/HGAL/MGC40441
Gene Symbols GCET2
Host Species Mouse
Immunogen GCET2 (NP_689998.1, 1 a.a. ∼ 178 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257144
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.