Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

glycine cleavage system protein H (aminomethyl carrier), Mouse, Clone: 3D8-A12, Abnova™

Catalog No. 89000683 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-683 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-683 Supplier Abnova Corporation Supplier No. H00002653M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant GCSH.

The enzyme system for cleavage of glycine (glycine cleavage system; EC 2.1.2.10), which is confined to the mitochondria, is composed of 4 protein components: P protein (a pyridoxal phosphate-dependent glycine decarboxylase; MIM 238300), H protein (a lipoic acid-containing protein), T protein (a tetrahydrofolate-requiring enzyme; MIM 238310), and L protein (a lipoamide dehydrogenase; MIM 238331). Glycine encephalopathy (GCE; MIM 605899), also called nonketotic hyperglycinemia (NKH), may be due to a defect in any one of these enzymes.[supplied by OMIM

Sequence: MALRVVRSVRALLCTLRAVPLPAAPCPPRPWQLGVGAVRTLRTGPALLSVRKFTEKHEWVTTENGIGTVGISNFAQEALGDVVYCSLPEVGTKLNKQDEFGALESVKAASELYSPLSGEVTEINEALAENPGLVNKSCYEDGWLIKMTLSNPSELDELMSEEAYEKYIKSIEE

Specifications

Antigen glycine cleavage system protein H (aminomethyl carrier)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3D8-A12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant GCSH.
Formulation PBS with no preservative; pH 7.4
Gene GCSH
Gene Accession No. BC000790
Gene Alias GCE/NKH
Gene Symbols GCSH
Host Species Mouse
Immunogen GCSH (AAH00790.1, 1 a.a. ∼ 173 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 2653
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.