Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GDF-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5754
This item is not returnable.
View return policy
Description
GDF-3 Monoclonal specifically detects GDF-3 in Human samples. It is validated for Immunocytochemistry.Specifications
GDF-3 | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
Q9NR23 | |
GDF3 | |
Chinese hamster ovary cell line CHO-derived recombinant human GDF-3 Ala251-Gly364 Accession # Q9NR23 | |
100 μg | |
Primary | |
Detects human GDF-3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) GDF-1, rhBMP-2, or rhBMP-6 is observed and approximately 5% cross-reactivity with recombinant mouse GDF-3 homodimer is observed. | |
Human | |
Purified |
Immunocytochemistry | |
741614 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
GDF3, GDF-3, growth differentiation factor 3, growth/differentiation factor 3, KFS3, MCOP7, MCOPCB6, Vgr-2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9573 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction