Learn More
growth differentiation factor 10, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant GDF10.
Supplier: Abnova Corporation H00002662A01
Description
The protein encoded by this gene is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in mice suggest that the protein encoded by this gene plays a role in skeletal morphogenesis. [provided by RefSeq
Sequence: MALAPPPRGLWQAKDISPIVKAARRDGELLLSAQLDSEERDPGVPRPSPYAPYILVYANDLAISEPNSVAVTLQRYDPFPAGDPEPRAAPNNSADPRVRRAAQATGPLQDNELPGLDERPPRAHAQHFHKHQLWPSPFRALKPRPGRKDRRKKGQEVFMAASQVLDFDEKTMQKARRKQWDEPRVCSRRYLKVDFADIGWNEWIISPKSFDAYYCAGACEFPMPKIVRPSNHATIQSIVRAVGIIPGIPEPCCVPDKMNSLGVLFLDENRNVVLKVYPNMSVDTCACRSpecifications
growth differentiation factor 10 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length recombinant GDF10. | |
GDF10 | |
BMP-3b/BMP3B | |
Mouse | |
50 μL | |
Cytokines | |
Primary | |
Human, Mouse | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
BC028237 | |
GDF10 | |
GDF10 (AAH28237, 1 a.a. ∼ 288 a.a) full-length recombinant protein with GST tag. | |
RUO | |
Yes | |
2662 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.