Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

growth differentiation factor 11, Mouse, Clone: 3G6, Abnova™

Catalog No. 89001169 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-169 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-169 Supplier Abnova Corporation Supplier No. H00010220M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GDF11.

The protein encoded by this gene is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in mice and Xenopus suggest that this protein is involved in mesodermal formation and neurogenesis during embryonic development. [provided by RefSeq

Sequence: ESRCCRYPLTVDFEAFGWDWIIAPKRYKANYCSGQCEYMFMQKYPHTHLVQQANPRGSAGPCCTPTKMSPINMLYFNDKQQIIYGKIPGMVVDRCGCS

Specifications

Antigen growth differentiation factor 11
Applications ELISA, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone 3G6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GDF11.
Formulation PBS with no preservative; pH 7.4
Gene GDF11
Gene Accession No. NM_005811
Gene Alias BMP-11/BMP11
Gene Symbols GDF11
Host Species Mouse
Immunogen GDF11 (NP_005802, 310 a.a. ∼ 407 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cytokines
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 10220
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.