Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gdown1 Sheep anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABE23MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE23MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE23MI Supplier MilliporeSigma Supplier No. ABE23
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Use Anti-Gdown1 Antibody (Sheep Polyclonal Antibody) validated in western blot, Chimmunoprecipitation to detect Gdown1 also known as Myocardial zonula adherens protein, GRINL1A upstream protein, Gup.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gdown1
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified sheep polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols MYZAP; MYOZAP
Host Species Sheep
Immunogen Histidine-tagged recombinant protein corresponding to human Gdown1.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_001018101
Target Species Human
Content And Storage 2-8°C for 12 months
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.