Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gemin2 Mouse anti-Human, Mouse, Clone: 2E17, MilliporeSigma™

Catalog No. 051540MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-154-0MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-154-0MI Supplier MilliporeSigma Supplier No. 051540
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Gemin2 Antibody, clone 2E17 is a Mouse Monoclonal Antibody for detection of Gemin2 also known as SMN interacting protein 1-delta and has been validated in western blot, immunocytochemistry, ELISA and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gemin2
Applications ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2E17
Conjugate Unconjugated
Formulation Purified mouse IgG1κ supernatant in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide
Gene Accession No. NM_001009182.1
Gene Symbols SIP1, SIP1-delta
Host Species Mouse
Immunogen His-tagged recombinant protein corresponding to human Gemin-2.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.