Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gem (nuclear organelle) associated protein 7, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018752 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-752 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-752 Supplier Abnova Corporation Supplier No. H00079760D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human GEMIN7 protein.

The protein encoded by this gene is a component of the core SMN complex, which is required for pre-mRNA splicing in the nucleus. The encoded protein is found in the nucleoplasm, in nuclear gems (Gemini of Cajal bodies), and in the cytoplasm. Three transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]

Sequence: MQTPVNIPVPVLRLPRGPDGFSRGFAPDGRRAPLRPEVPEIQECPIAQESLESQEQRARAALRERYLRSLLAMVGHQVSFTLHEGVRVAAHFGATDLDVANFYVSQLQTPIGVQAEALLRCSDIISYTFKP

Specifications

Antigen gem (nuclear organelle) associated protein 7
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human GEMIN7 protein.
Formulation PBS with no preservative; pH 7.4
Gene GEMIN7
Gene Accession No. NM_001007269.1
Gene Alias SIP3
Gene Symbols GEMIN7
Host Species Rabbit
Immunogen GEMIN7 (NP_001007270.1, 1 a.a. ∼ 131 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79760
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.