Learn More
Anti- Gemtuzumab (Anti-Idiotype) Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Gemtuzumab |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 975155 |
| Conjugate | Alexa Fluor 594 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Host Species | Mouse |
| Immunogen | Full length Gemtuzumab |
| Purification Method | Protein A or G purified from hybridoma culture supernatant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.