Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gephyrin Mouse, Unlabeled, Clone: 45, BD

Catalog No. BDB610585 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610585 150 μg
BDB610584 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610585 Supplier BD Biosciences Supplier No. 610585
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The sub-membraneous region of the postsynaptic neuron is an intricate network of cytoskeletal elements generally known as the postsynaptic density. It is thought that this elaborate cytoskeletal region is critical for receptor targeting, clustering, and efficient signal input. Gephyrin, a 93 kDa protein, was identified as a result of its ability to bind to polymerized tubulin (microtubules). Although expressed in all tissues, gephyrin mRNA is found most abundantly in brain. Alternative splicing produces at least five different mRNAs. Gephyrin colocalizes and interacts with the glycine receptor at the postsynaptic density. It is possible that Gephyrin acts as an anchor between the glycine receptor and microtubules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gephyrin
Applications Western Blot
Classification Monoclonal
Clone 45
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat Gephyrin aa. 569-726
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.