Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFAP Mouse, Unlabeled, Clone: 4A11, BD

Catalog No. BDB556327 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556327 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556327 Supplier BD Biosciences Supplier No. 556327
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GFAP (Glial Fibrillary Acid Protein) is the major protein of glial filaments in differentiated astrocytes. BD Biosciences Pharmingen offers a panel of monoclonal antibodies (4A11, 1B4, 2E1) that specifically recognize GFAP. They do not cross-react with other intermediate filaments such as vimentin, neurofilament proteins, desmin, keratin, neurotubules or microfilaments. Clone 4A11 is specific for GFAP. Bovine spinal cord homogenate was used as immunogen. This antibody has broad species reactivity, recognizing GFAP in brain homogenates from human, mouse, rat, bovine, ovine, canine, porcine, rabbit, guinea pig and chicken. 4A11 is particularly useful for identifying GFAP in immunohistochemistry of frozen and formalin-fixed, paraffin-embedded brain tissue sections. Additional applications include western blot analysis and indirect immunofluorescence of tissue-cultured cells. BD Biosciences Pharmingen offers additional GFAP specific antibodies: clone 2E1 (Mfr. No. 556329), 1B4 (Mfr. No. 556328), and clones 1B4, 4A11, 2E1 combined and available as a “cocktail” (Mfr. No. 556330).

Host Species: Mouse
Clone: 4A11
Isotype: IgG2b, κ
Species Reactivity: Human
Immunogen: Cow spinal cord homogenate

Immunofluorescence, Immunohistochemistry, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen GFAP
Applications Immunohistochemistry
Classification Monoclonal
Clone 4A11
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Cow spinal cord homogenate
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Bovine, Canine, Chicken, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Sheep
Content And Storage Store undiluted at 4°C
Isotype IgG2b
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.