Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP Rabbit anti-All, Polyclonal, MilliporeSigma™

Catalog No. AB10145MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB10145MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB10145MI Supplier MilliporeSigma Supplier No. AB10145
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-GFP Antibody is validated for use in western blotting for the detection of GFP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit immunoglobulin in buffer containing PBS containing 0.05% sodium azide.
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the full length of GFP protein.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) AAA27722
Target Species All species
Content And Storage 2-8°C for 12 months in undiluted aliquotes Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.