Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP (Green Fluorescent Protein) Chicken anti-Vertebrates, Polyclonal, MilliporeSigma™

Catalog No. 06896MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-896-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-896-MI Supplier MilliporeSigma Supplier No. 06896
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Anti-GFP (Green Fluorescent Protein) Antibody detects level of GFP (Green Fluorescent Protein) and has been published and validated for use in IC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFP
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified chicken polyclonal IgY from egg yolk in buffer containing 70% storage buffer (0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide) and 30% glycerol. Store at −20°C.
Gene Symbols GFP
Host Species Chicken
Immunogen His-tagged green fluorescent protein of Aequorea victoria.
Purification Method Ammonium Sulfate Precipitation and PEG
Quantity 200 μg
Primary or Secondary Primary
Target Species Vertebrates
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Purified
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.